
Pentafluorophenyl-1-methanesulphonate
Catalog # APOSPC10572-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:76 °C/0.6mmHg
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:161912-36-3
Formula:C₇H₃O₃F₅S
MDL Number:MFCD06659544