
2-Methoxy-5-(trifluoromethoxy)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:145742-65-0
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD04115958
:220.15 g/mol
Density:1.36 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together