2-Chloro-2,2-difluoro-1-(4-hydroxypiperidino)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:478258-79-6
:
Formula:C₇H₁₀ClF₂NO₂
MDL Number:MFCD02082432
:213.61 g/mol
Boiling Pt:117 °C/1mmHg
Storage Temperature:Ambient
Frequently Bought Together