Order Entry
Italy
ContactUsLinkComponent
5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole 97%
Irritant
5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole 97%
Catalog # APOSPC1029-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole 97%
Catalog # APOSPC1029-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :192.19 g/mol
Melting Pt:178…180 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:150433-22-0
Formula:C₈H₁₁F₃N₂
MDL Number:MFCD00729044