
3-Methoxy-4-{[3-(trifluoromethyl)benzyl]oxy}benzenecarbaldehyde
Catalog # APOSPC10256-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:310.28 g/mol
Melting Pt:64…65 °C
:GHS07
Storage Temperature:Ambient
CAS Number:380427-19-0
Formula:C₁₆H₁₃F₃O₃
MDL Number:MFCD02255725