
2-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:336880-01-4
:
Formula:C₁₄H₁₀ClFO₂
MDL Number:MFCD01590675
Melting Pt:71…73 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together