Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
APOSPC10243-1G
:
Apollo Scientific
:
N-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetamide
APOSPC10243-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
245.26 g/mol
:
139…141 °C
:
GHS07
:
Ambient
:
131179-72-1
:
C₁₄H₁₂FNO₂
:
MFCD03425776