3-Methoxy-3-oxo-1-propenyl-4-(trifluoromethyl)benzenecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:400878-21-9
:
Formula:C₁₂H₉F₃O₄
MDL Number:MFCD03001265
:274.2 g/mol
Melting Pt:86…88 °C
Storage Temperature:Ambient
Frequently Bought Together