Order Entry
Italy
ContactUsLinkComponent
Aluminium fluoride trihydrate 97%
Irritant
Aluminium fluoride trihydrate 97%
Catalog # APOSPC1009-500G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Aluminium fluoride trihydrate 97%
Catalog # APOSPC1009-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :138.02 g/mol
Density:2.88 g/cm³ (25 °C)
EINECS:232-051-1
 :GHS07
Storage Temperature:Ambient
CAS Number:15098-87-0
Formula:AlF₃·3H₂O
MDL Number:MFCD00149141