
2,2-Dichloro-1,1,1,4,4,4-hexafluorobutane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:162462-08-0
:
Formula:C₄H₂Cl₂F₆
MDL Number:MFCD01320732
:234.96 g/mol
Boiling Pt:75…76 °C
:GHS07
Storage Temperature:Ambiente
Frequently Bought Together