
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:129946-88-9
:
Formula:C₁₄H₈F₆O₃S₂
MDL Number:MFCD00236132
:402.325 g/mol
Melting Pt:150 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together