Order Entry
Italy
ContactUsLinkComponent
2-Nitro-4-(trifluoromethyl)benzoyl chloride 98%
Corrosive
2-Nitro-4-(trifluoromethyl)benzoyl chloride 98%
Catalog # APOSPC01532-10G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Nitro-4-(trifluoromethyl)benzoyl chloride 98%
Catalog # APOSPC01532-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :253.57 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:81108-81-8
Formula:C₈H₃ClF₃NO₃
MDL Number:MFCD07777203