Order Entry
Italy
ContactUsLinkComponent
4,5-Difluoro-2-methylbenzimidazole
Irritant
4,5-Difluoro-2-methylbenzimidazole
Catalog # APOSPC0115-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,5-Difluoro-2-methylbenzimidazole
Catalog # APOSPC0115-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :168.15 g/mol
Melting Pt:250 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:229957-09-9
Formula:C₈H₆F₂N₂
MDL Number:MFCD00665871