
Ethyl-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2549-19-1
:
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD00139509
:204.2300 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together