
p-Toluenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98-59-9
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00007450
:190.65 g/mol
Boiling Pt:146 °C/15mmHg
Melting Pt:67…70 °C
Density:1.35 g/cm³ (25 °C)
Flash Pt:128 °C
Storage Temperature:Ambient
EINECS:202-684-8
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together