1-(4-Chlorophenyl)propan-1-amine 95%
Catalog # APOSOR964966-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:169.66 g/mol
Storage Temperature:Ambient
CAS Number:74788-46-8
Formula:C₉H₁₂ClN
MDL Number:MFCD08691240