Order Entry
Italy
ContactUsLinkComponent
EDTA disodium salt 98%
EDTA disodium salt 98%
Catalog # APOSOR964961-1KG
Supplier:  Apollo Scientific
CAS Number:  
EDTA disodium salt 98%
Catalog # APOSOR964961-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672