5-Bromo-2-formyl-3-methoxypyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1087659-24-2
:
Formula:C₇H₆BrNO₂
MDL Number:MFCD11857632
:216.0300 g/mol
Melting Pt:70…80 °C
Storage Temperature:Refrigerator
Frequently Bought Together