N,N-Diethyl-4-nitroaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2216-15-1
:
Formula:C₁₀H₁₄N₂O₂
MDL Number:MFCD00043606
:194.23 g/mol
Melting Pt:77.5 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
Frequently Bought Together