2,3-Dihydrobenzo[1,4]dioxine-5-carbonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38871-41-9
:
Formula:C₉H₇ClO₃
MDL Number:MFCD04972616
:198.61 g/mol
Storage Temperature:Ambiente
Frequently Bought Together