Order Entry
Italy
ContactUsLinkComponent
Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Irritant
Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Catalog # APOSOR960335-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-7-aminothieno[2,3-b]pyrazine-6-carboxylate
Catalog # APOSOR960335-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.25 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:56881-21-1
Formula:C₉H₉N₃O₂S
MDL Number:MFCD00096361