Ethyl-4-formyl-1H-pyrazole-5-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:179692-09-2
:MFCD07643185
Formula:C₇H₈N₂O₃
MDL Number:MFCD03768340
:168.15 g/mol
Storage Temperature:Ambient
Frequently Bought Together