Order Entry
Italy
ContactUsLinkComponent
2-Methoxy-5-methylnicotinonitrile 98%
2-Methoxy-5-methylnicotinonitrile 98%
Catalog # APOSOR953188-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-5-methylnicotinonitrile 98%
Catalog # APOSOR953188-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :148.17 g/mol
Storage Temperature:Ambient
CAS Number:149379-73-7
Formula:C₈H₈N₂O
MDL Number:MFCD13563068