Order Entry
Italy
ContactUsLinkComponent
 
Ethyl-4-aminopyrimidine-5-carboxylate 97%
Catalog # APOSOR953105-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-4-aminopyrimidine-5-carboxylate 97%
Catalog # APOSOR953105-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.17 g/mol
Storage Temperature:Ambient
CAS Number:65195-35-9
Formula:C₇H₉N₃O₂
MDL Number:MFCD01028678