Order Entry
Italy
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 98%
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 98%
Catalog # APOSOR952909-5G
Supplier:  Apollo Scientific
CAS Number:  
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 98%
Catalog # APOSOR952909-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.19 g/mol
Storage Temperature:Ambient
CAS Number:530-93-8
Formula:C₁₀H₁₀O
MDL Number:MFCD00001727