Order Entry
Italy
ContactUsLinkComponent
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid
Irritant
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSOR9488-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid
Catalog # APOSOR9488-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :176.1700 g/mol
Melting Pt:147…149 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:21801-79-6
Formula:C₉H₈N₂O₂
MDL Number:MFCD00244615