Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Nitro-2-picoline

4-Nitro-2-picoline

Irritant
Supplier:   Apollo Scientific
4-Nitro-2-picoline
Supplier:  Apollo Scientific
Certificates
4-Nitro-2-picoline

2 Options Available Below

Product Details & Documents
Synonyms:  2-Methyl-4-nitropyridine
  • CAS Number:13508-96-8
  • Formula:C₆H₆N₂O₂
  • MDL Number:MFCD03095077
  • Molecular Weight:138.1200 g/mol
  • Melting Pt:42…44 °C
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.