Order Entry
Italy
ContactUsLinkComponent
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
Irritant
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
Catalog # APOSOR9436-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98%
Catalog # APOSOR9436-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:135…137 °C
 :113.1600 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:75885-58-4
Formula:C₆H₁₁NO
MDL Number:MFCD00216614