Isopropenyloxytrimethylsilane 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1833-53-0
:
Formula:C₆H₁₄OSi
MDL Number:MFCD00042845
:130.26 g/mol
Storage Temperature:Ambient
Frequently Bought Together
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
Water ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
1,2-Dimethoxyethane, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND