1,3-Dimethyl-1H-pyrazole-4-sulphonyl chloride 95%
Catalog # APOSOR938453-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:194.64 g/mol
Storage Temperature:Ambient
CAS Number:89501-93-9
Formula:C₅H₇ClN₂O₂S
MDL Number:MFCD03944999