1,2-Di(2-pyridyl)ethylene 95%
Catalog # APOSOR936887-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.22 g/mol
Storage Temperature:Ambient
CAS Number:1437-15-6
Formula:C₁₂H₁₀N₂
MDL Number:MFCD00006356