4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) 98%
Catalog # APOSOR936587-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:158.20 g/mol
Storage Temperature:Ambient
CAS Number:13380-84-2
Formula:C₈H₁₄O₃
MDL Number:MFCD01734677