4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19438-60-9
:
Formula:C₉H₁₂O₃
MDL Number:MFCD00005927
:168.19 g/mol
Storage Temperature:Ambient
Frequently Bought Together