[1-(2-Methyl-1,3-thiazol-4-yl)ethyl]amine dihydrochloride 95%
Catalog # APOSOR931075-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.15 g/mol
Storage Temperature:Ambient
CAS Number:317830-81-2
Formula:C₆H₁₂Cl₂N₂S
MDL Number:MFCD07781067