2-Methyl-4,6-bis[(n-octylthio)methyl]phenol 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Methyl-4,6-bis(n-octylsulphanylmethyl)phenol, 2-Methyl-4,6-bis((octylthio)methyl)phenol
CAS Number:110553-27-0
:
Formula:C₂₅H₄₄OS₂
MDL Number:MFCD00134699
:424.75 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together