Order Entry
Italy
ContactUsLinkComponent
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 95%
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 95%
Catalog # APOSOR928753-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 95%
Catalog # APOSOR928753-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.6 g/mol
Storage Temperature:Ambient
CAS Number:871024-38-3
Formula:C₇H₆ClN₃
MDL Number:MFCD12924466