3-Isopropyl-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952182-05-7
:
Formula:C₈H₁₄N₄
MDL Number:MFCD09743507
:166.23 g/mol
Storage Temperature:Ambient
Frequently Bought Together