Order Entry
Italy
Orders LinkContactUsLinkComponent
2,6-Dimethoxynaphthalene ≥98%

2,6-Dimethoxynaphthalene ≥98%

New Product
Nome del fornitore:   Apollo Scientific
2,6-Dimethoxynaphthalene ≥98%
Nome del fornitore:  Apollo Scientific
New Product
Certificates
2,6-Dimethoxynaphthalene ≥98%

6 Options Available Below

Product Details & Documents
  • CAS Number:5486-55-5
  • Formula:C₁₂H₁₂O₂
  • MDL Number:MFCD00004063
  • Molecular Weight:188.23 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Disponibilità
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 39,10 €
    39,10 €
    1 * 5 g
       39,10 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 151,00 €
    151,00 €
    1 * 25 g
       151,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 601,00 €
    601,00 €
    1 * 100 g
       601,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2 950,00 €
    2 950,00 €
    1 * 500 g
       2 950,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.