
1,5-Dimethyl-1H-pyrazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31728-75-3
:
Formula:C₆H₈N₂O₂
MDL Number:MFCD02090876
:140.14 g/mol
Melting Pt:181…183 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together