2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65894-83-9
:
Formula:C₉H₁₇NS
MDL Number:MFCD01680822
:171.30 g/mol
Storage Temperature:Ambient
Frequently Bought Together