Chloride ionophore I 97%
Catalog # APOSOR918293-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:703.11 g/mol
Storage Temperature:Ambient
CAS Number:32195-55-4
Formula:C₄₄H₂₈ClMnN₄
MDL Number:MFCD00012153