Order Entry
Italy
ContactUsLinkComponent
2-(4-Chlorophenyl)-2-methylpropanol 98%
2-(4-Chlorophenyl)-2-methylpropanol 98%
Catalog # APOSOR917141-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chlorophenyl)-2-methylpropanol 98%
Catalog # APOSOR917141-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :184.66 g/mol
Storage Temperature:Ambient
CAS Number:80854-14-4
Formula:C₁₀H₁₃ClO
MDL Number:MFCD06657569