
1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:864068-96-2
:
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD08271941
:186.2100 g/mol
Melting Pt:95…96.5 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together