3-(Hydroxymethyl)-2-hydroxynaphthalene 98%
Catalog # APOSOR914542-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:174.195 g/mol
Storage Temperature:Ambient
CAS Number:30159-70-7
Formula:C₁₁H₁₀O₂
MDL Number:MFCD11504950