
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5617-70-9
:
Formula:C₈H₁₀O₄
MDL Number:MFCD00042796
:170.16 g/mol
Melting Pt:69…73 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together