Order Entry
Italy
ContactUsLinkComponent
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%
Catalog # APOSOR913301-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Benzo[d]oxazol-2-yl)-2-methylpropanenitrile 95%
Catalog # APOSOR913301-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.2100 g/mol
Storage Temperature:Ambient
CAS Number:157763-81-0
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD08166396