Ethyl-5-formyl-1H-pyrrole-2-carboxylate 95%
Catalog # APOSOR912411-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.1600 g/mol
Storage Temperature:Ambient
CAS Number:7126-50-3
Formula:C₈H₉NO₃
MDL Number:MFCD00159462