Order Entry
Italy
ContactUsLinkComponent
((R)-Piperidin-2-yl)methanol 95%
((R)-Piperidin-2-yl)methanol 95%
Catalog # APOSOR910571-1G
Supplier:  Apollo Scientific
CAS Number:  
((R)-Piperidin-2-yl)methanol 95%
Catalog # APOSOR910571-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :115.18 g/mol
Storage Temperature:Ambient
CAS Number:3197-44-2
Formula:C₆H₁₃NO
MDL Number:MFCD11036292