Order Entry
Italy
ContactUsLinkComponent
Methyl-3-bromo-4-hydroxybenzoate 98%
Irritant
Methyl-3-bromo-4-hydroxybenzoate 98%
Catalog # APOSOR908176-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-3-bromo-4-hydroxybenzoate 98%
Catalog # APOSOR908176-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :231.05 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:29415-97-2
Formula:C₈H₇BrO₃
MDL Number:MFCD06203850