
4-Morpholin-4-yl-1,2,5-thiadiazol-3-ol
Catalog # APOSOR9072-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.22 g/mol
Melting Pt:196…200 °C
:GHS07
Storage Temperature:Ambient
CAS Number:30165-97-0
Formula:C₆H₉N₃O₂S
MDL Number:MFCD01820806